5-Amino-2-methoxypyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024176-10G 024176-50G
FLUO024176-10GEA 0 GBP
FLUO024176-10G FLUO024176-50G
5-Amino-2-methoxypyridine
5-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 265
Melting Pt: 28…30 °C
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR