5-Bromo-2-chloropyrimidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023429-1G 023429-5G
FLUO023429-1GEA 0 GBP
FLUO023429-1G FLUO023429-5G
5-Bromo-2-chloropyrimidine
5-Bromo-2-chloropyrimidine
Formula: C₄H₂BrClN₂
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR