5-Chloro-o-anisidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chloro-2-methoxyaniline, 5-Chloro-2-anisidine, 2-Amino-4-chloroanisole
020202-25G
020202-100G
020202-500G
FLUO020202-25GEA
0
GBP
FLUO020202-25G
FLUO020202-100G
FLUO020202-500G
5-Chloro-o-anisidine
5-Chloro-o-anisidine
Formula:
C₇H₈ClNO MW: 157.6 g/mol Boiling Pt: 259 °C (1013 hPa) Melting Pt: 84 °C |
MDL Number:
MFCD00007777 CAS Number: 95-03-4 EINECS: 202-385-2 UN: 2233 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...