6-Aminonicotinonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

020194-1G 020194-25G 020194-5G
FLUO020194-1GEA 0 GBP
FLUO020194-1G FLUO020194-25G FLUO020194-5G
6-Aminonicotinonitrile
6-Aminonicotinonitrile
Formula: C₆H₅N₃
Boiling Pt: 240…250 °C (15 mmHg)
Melting Pt: 164 °C
MDL Number: MFCD00128902
CAS Number: 4214-73-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR