1-(2-(Trifluoromethyl)phenyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

020167-1G 020167-5G
FLUO020167-1GEA 0 GBP
FLUO020167-1G FLUO020167-5G
1-(2-(Trifluoromethyl)phenyl)imidazole
1-(2-(Trifluoromethyl)phenyl)imidazole
Formula: C₁₀H₇F₃N₂
Boiling Pt: 75 °C (0,025 mmHg)
Melting Pt: 50…53 °C
MDL Number: MFCD00041206
CAS Number: 25371-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR