4-Iodopyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019463-1G
019463-25G
019463-5G
FLUO019463-1GEA
0
GBP
FLUO019463-1G
FLUO019463-25G
FLUO019463-5G
4-Iodopyridine
4-Iodopyridine
Formula:
C₅H₄IN |
MDL Number:
MFCD02093937 CAS Number: 15854-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...