2-Fluoro-N-methylaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019280-10G
019280-25G
FLUO019280-10GEA
0
GBP
FLUO019280-10G
FLUO019280-25G
2-Fluoro-N-methylaniline
2-Fluoro-N-methylaniline
Formula:
C₇H₈FN |
MDL Number:
MFCD00028030 CAS Number: 1978-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...