2,6-Dimethylbenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019244-5G 019244-1G
FLUO019244-5GEA 0 GBP
FLUO019244-5G FLUO019244-1G
2,6-Dimethylbenzenesulphonyl chloride
2,6-Dimethylbenzenesulphonyl chloride
Formula: C₈H₉ClO₂S
MDL Number: MFCD03789123
CAS Number: 2905-29-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR