N-Methylphthalimide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methylisoindoline-1,3-dione

018866-10G 018866-25G
FLUO018866-10GEA 0 GBP
FLUO018866-10G FLUO018866-25G
N-Methylphthalimide
N-Methylphthalimide
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 286
Melting Pt: 134
MDL Number: MFCD00023063
CAS Number: 550-44-7
EINECS: 208-982-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR