3-(p-Tolyl)propionic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018857.10G
018857-1G
018857-5G
FLUO018857.10GEA
0
GBP
FLUO018857.10G
FLUO018857-1G
FLUO018857-5G
3-(p-Tolyl)propionic acid
3-(p-Tolyl)propionic acid
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD00016561 CAS Number: 1505-50-6 EINECS: 216-132-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...