1-(3-Methoxyphenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018809-10G 018809-1G
FLUO018809-10GEA 0 GBP
FLUO018809-10G FLUO018809-1G
1-(3-Methoxyphenyl)-2-thiourea
1-(3-Methoxyphenyl)-2-thiourea
Formula: (C₆H₄(OCH₃))NC(S)NH₂
MDL Number: MFCD00060446
CAS Number: 37014-08-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR