Isopropyl anthranilate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018753-10G 018753-25G 018753-5G
FLUO018753-10GEA 0 GBP
FLUO018753-10G FLUO018753-25G FLUO018753-5G
Isopropyl anthranilate
Isopropyl anthranilate
Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00060604
CAS Number: 18189-02-1
EINECS: 242-074-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR