4-Chloro-2-iodoanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018381-25G 018381-5G
FLUO018381-25GEA 0 GBP
FLUO018381-25G FLUO018381-5G
4-Chloro-2-iodoanisole
4-Chloro-2-iodoanisole
Formula: C₇H₆ClIO
MDL Number: MFCD00052858
CAS Number: 52807-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR