1-(3-Bromophenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

018318-1G 018318-5G
FLUO018318-1GEA 0 GBP
FLUO018318-1G FLUO018318-5G
1-(3-Bromophenyl)-2-thiourea
1-(3-Bromophenyl)-2-thiourea
Formula: (C₆H₄Br)NHC(S)NH₂
MDL Number: MFCD00060440
CAS Number: 21327-14-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR