3-(4-Bromophenyl)propionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018312-1G 018312-5G
FLUO018312-1GEA 0 GBP
FLUO018312-1G FLUO018312-5G
3-(4-Bromophenyl)propionic acid
3-(4-Bromophenyl)propionic acid
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD01310793
CAS Number: 1643-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR