N-(8-Bromooctyl)phthalimide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018143.25G 018143-1G 018143-5G
FLUO018143.25GEA 1050 GBP
FLUO018143.25G FLUO018143-1G FLUO018143-5G
N-(8-Bromooctyl)phthalimide
N-(8-Bromooctyl)phthalimide
Formula: C₁₆H₂₀BrNO₂
MDL Number: MFCD00060523
CAS Number: 17702-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR