N-(10-Bromodecyl)phthalimide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018023-5G 018023-25G
FLUO018023-5GEA 0 GBP
FLUO018023-5G FLUO018023-25G
N-(10-Bromodecyl)phthalimide
N-(10-Bromodecyl)phthalimide
Formula: C₁₈H₂₄BrNO₂
MDL Number: MFCD00060521
CAS Number: 24566-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR