3-Amino-2,6-dichloropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine

017208-5G 017208-1G
FLUO017208-5GEA 0 GBP
FLUO017208-5G FLUO017208-1G
3-Amino-2,6-dichloropyridine
3-Amino-2,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 305
Melting Pt: 119
Density: 1.497 g/cm³ (20 °C)
MDL Number: MFCD00023417
CAS Number: 62476-56-6
EINECS: 263-559-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR