4-(Trifluoromethyl)anilinium chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Aminobenzotrifluoride HCl, 4-Aminobenzotrifluoride hydrochloride, α,α,α-Trifluoro-p-toluidine HCl, α,α,α-Trifluoro-p-toluidine hydrochloride, 4-(Trifluoromethyl)aniline hydrochloride

013906-5G 013906-25G 013906-100G
FLUO013906-5GEA 0 GBP
FLUO013906-5G FLUO013906-25G FLUO013906-100G
4-(Trifluoromethyl)anilinium chloride
4-(Trifluoromethyl)anilinium chloride
Formula: C₇H₇ClF₃N
Melting Pt: 200 °C
MDL Number: MFCD00007897
CAS Number: 90774-69-9
EINECS: 292-679-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR