2,6-Dichloro-3-(trifluoromethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013884-5G
013884-1G
FLUO013884-5GEA
0
GBP
FLUO013884-5G
FLUO013884-1G
2,6-Dichloro-3-(trifluoromethyl)pyridine
2,6-Dichloro-3-(trifluoromethyl)pyridine
Formula:
C₆H₂Cl₂F₃N Boiling Pt: 195 °C (1013 hPa) |
MDL Number:
MFCD00042245 CAS Number: 55304-75-1 EINECS: 259-585-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...