Tetrafluorophthalic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013867-25G
013867-5G
FLUO013867-25GEA
0
GBP
FLUO013867-25G
FLUO013867-5G
Tetrafluorophthalic anhydride
Tetrafluorophthalic anhydride
Formula:
C₈F₄O₃ |
MDL Number:
MFCD00039697 CAS Number: 652-12-0 EINECS: 211-485-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...