3-Iodo-p-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013858-25G 013858-100G
FLUO013858-25GEA 0 GBP
FLUO013858-25G FLUO013858-100G
3-Iodo-p-toluic acid
3-Iodo-p-toluic acid
Formula: C₈H₇IO₂
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 210…211 °C
MDL Number: MFCD00010392
CAS Number: 82998-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR