2-Bromo-5-(trifluoromethyl)benzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013790-25G
013790-5G
FLUO013790-25GEA
0
GBP
FLUO013790-25G
FLUO013790-5G
2-Bromo-5-(trifluoromethyl)benzaldehyde
2-Bromo-5-(trifluoromethyl)benzaldehyde
Formula:
C₈H₄BrF₃O MW: 253.02 g/mol |
MDL Number:
MFCD04973760 CAS Number: 102684-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...