5-Bromo-2,4-dichloropyrimidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011581-5G 011581-100G 011581-25G
FLUO011581-5GEA 0 GBP
FLUO011581-5G FLUO011581-100G FLUO011581-25G
5-Bromo-2,4-dichloropyrimidine
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.781 g/cm³ (20 °C)
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR