(3,4-Xylyloxy)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011161-25G 011161-100G
FLUO011161-25GEA 0 GBP
FLUO011161-25G FLUO011161-100G
(3,4-Xylyloxy)acetic acid
(3,4-Xylyloxy)acetic acid
Formula: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180.2 g/mol
MDL Number: MFCD00017595
CAS Number: 13335-73-4
EINECS: 236-386-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR