3-Nitrophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

010815-25G 010815-5G
FLUO010815-25GEA 0 GBP
FLUO010815-25G FLUO010815-5G
3-Nitrophenylboronic acid
3-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 290…291 °C
MDL Number: MFCD00007193
CAS Number: 13331-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR