4-(2-Aminoethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-Pyridyl)ethylamine, 2-Pyridin-4-yl-ethylamine

010690-1G 010690-5G 010690-25G
FLUO010690-1GEA 0 GBP
FLUO010690-1G FLUO010690-5G FLUO010690-25G
4-(2-Aminoethyl)pyridine
4-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 231
Melting Pt: 76
Density: 1.0238 g/cm³ (25 °C)
MDL Number: MFCD00038045
CAS Number: 13258-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR