2-Fluoro-4-iodoaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010682-25G 010682-100G
FLUO010682-25GEA 0 GBP
FLUO010682-25G FLUO010682-100G
2-Fluoro-4-iodoaniline
Formula: C₆H₅FIN
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 52…54 °C
MDL Number: MFCD00011738
CAS Number: 29632-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR