3-Fluoro-p-anisidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-4-anisidine, 3-Fluoro-4-methoxyaniline, 4-Amino-2-fluoroanisole

010679-1G 010679-5G 010679 010679-25G
FLUO010679-1GEA 0 GBP
FLUO010679-1G FLUO010679-5G FLUO010679 FLUO010679-25G
3-Fluoro-p-anisidine
3-Fluoro-p-anisidine
Formula: C₇H₈FNO
MDL Number: MFCD00075040
CAS Number: 366-99-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR