2-Amino-5-bromo-3-nitropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

010399-1G 010399.5G 010399-25G
FLUO010399-1GEA 0 GBP
FLUO010399-1G FLUO010399.5G FLUO010399-25G
2-Amino-5-bromo-3-nitropyridine
2-Amino-5-bromo-3-nitropyridine
Formula: C₅H₄BrN₃O₂
Melting Pt: 215…216 °C
MDL Number: MFCD00047441
CAS Number: 6945-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR