3,5-Bis(trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010341-1G 010341-2.5G
FLUO010341-1GEA 0 GBP
FLUO010341-1G FLUO010341-2.5G
3,5-Bis(trifluoromethyl)pyridine
3,5-Bis(trifluoromethyl)pyridine
Formula: C₇H₃F₆N
MW: 215.1 g/mol
Boiling Pt: 117…118 °C (1013 hPa)
MDL Number: MFCD00128904
CAS Number: 20857-47-0
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR