1-(4-Pyridyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine

009853-1G 009853-5G
FLUO009853-1GEA 0 GBP
FLUO009853-1G FLUO009853-5G
1-(4-Pyridyl)piperazine
1-(4-Pyridyl)piperazine
Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Boiling Pt: 195…200 °C (0.3 mmHg)
Melting Pt: 140
MDL Number: MFCD00040745
CAS Number: 1008-91-9
EINECS: 213-764-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR