1-(2-Pyridyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(1-Piperazinyl)pyridine

009852-5G 009852-25G 009852-100G
FLUO009852-5GEA 0 GBP
FLUO009852-5G FLUO009852-25G FLUO009852-100G
1-(2-Pyridyl)piperazine
1-(2-Pyridyl)piperazine
Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 85…87 °C
Density: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00006216
CAS Number: 34803-66-2
EINECS: 252-220-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR