1-(3-Methylphenyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009808-1G 009808-5G
FLUO009808-1GEA 0 GBP
FLUO009808-1G FLUO009808-5G
1-(3-Methylphenyl)piperazine
1-(3-Methylphenyl)piperazine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00040734
CAS Number: 41186-03-2
EINECS: 255-251-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR