1-(2-Methylphenyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009807-5G 009807-25G 009807-100G
FLUO009807-5GEA 0 GBP
FLUO009807-5G FLUO009807-25G FLUO009807-100G
1-(2-Methylphenyl)piperazine
1-(2-Methylphenyl)piperazine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00040729
CAS Number: 39512-51-1
EINECS: 254-481-9
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR