2-Bromo-4-methylpyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009754-1G 009754-5G
FLUO009754-1GEA 0 GBP
FLUO009754-1G FLUO009754-5G
2-Bromo-4-methylpyridine
2-Bromo-4-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 42…44 °C (0,6 mmHg)
Melting Pt: 218…228 °C
Density: 1,545 g/cm³ (20 °C)
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR