2-(Aminomethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine
009750-100G
009750-25G
FLUO009750-100GEA
0
GBP
FLUO009750-100G
FLUO009750-25G
2-(Aminomethyl)pyridine
2-(Aminomethyl)pyridine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 197 Melting Pt: 81 Density: 1.0583 g/cm³ (20 °C) |
MDL Number:
MFCD00006360 CAS Number: 3731-51-9 EINECS: 223-090-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...