4-Fluoro-N-methylaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009651-10G 009651-25G 009651-5G
FLUO009651-10GEA 0 GBP
FLUO009651-10G FLUO009651-25G FLUO009651-5G
4-Fluoro-N-methylaniline
4-Fluoro-N-methylaniline
Formula: C₇H₈FN
MDL Number: MFCD00017942
CAS Number: 459-59-6
EINECS: 207-294-1
UN: 2511
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR