4-Chloro-N-methylaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009650-10G
009650-25G
009650-1G
009650-5G
FLUO009650-10GEA
0
GBP
FLUO009650-10G
FLUO009650-25G
FLUO009650-1G
FLUO009650-5G
4-Chloro-N-methylaniline
4-Chloro-N-methylaniline
Formula:
C₇H₈ClN Boiling Pt: 239 °C (1013 hPa) Density: 1.169 g/cm³ (12 °C) |
MDL Number:
MFCD00000614 CAS Number: 932-96-7 EINECS: 213-262-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...