2-Isobutylthiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009620-5G 009620-25G
FLUO009620-5GEA 0 GBP
FLUO009620-5G FLUO009620-25G
2-Isobutylthiazole
2-Isobutylthiazole
Formula: C₇H₁₁NS
MW: 141.24 g/mol
Boiling Pt: 180 °C (1013 hPa)
Density: 0,9953 g/cm³ (25 °C)
MDL Number: MFCD00005334
CAS Number: 18640-74-9
EINECS: 242-470-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR