2-(Trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009394-1G 009394-5G
FLUO009394-1GEA 0 GBP
FLUO009394-1G FLUO009394-5G
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
Boiling Pt: 138…139 °C (1013 hPa)
MDL Number: MFCD02183562
CAS Number: 368-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR