N-Methyl-N-(trimethylsilyl)trifluoroacetamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

009302-100G 009302-250G 009302-25G
FLUO009302-100GEA 0 GBP
FLUO009302-100G FLUO009302-250G FLUO009302-25G
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formula: C₆H₁₂F₃NOSi
Boiling Pt: 130 °C (1013 hPa)
Density: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR