1-(2-Fluorophenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (2-Fluorophenyl)thiourea, N-(2-Fluorophenyl)thiourea, 2-Fluorophenylthiourea

009242-25G 009242-5G
FLUO009242-25GEA 0 GBP
FLUO009242-25G FLUO009242-5G
1-(2-Fluorophenyl)-2-thiourea
1-(2-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 146 °C
Density: 1.423 g/cm³ (20 °C)
MDL Number: MFCD00041148
CAS Number: 656-32-6
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR