1-Ethyl-3-methylimidazoliumtetrafluoroborate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate
009180-1G
009180-5G
FLUO009180-1GEA
0
GBP
FLUO009180-1G
FLUO009180-5G
1-Ethyl-3-methylimidazoliumtetrafluoroborate
1-Ethyl-3-methylimidazolium tetrafluoroborate
Formula:
C₆H₁₁BF₄N₂ MW: 197.97 g/mol Boiling Pt: ∼200 °C (1013 hPa) Melting Pt: ∼15 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: >110 °C |
MDL Number:
MFCD00216668 CAS Number: 143314-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...