α,α,α-Trifluoro-2,5-diaminotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,5-Diaminobenzotrifluoride, 2-(Trifluoromethyl)benzene-1,4-diamine, 2-(Trifluoromethyl)-p-phenylenediamine, 2-(Trifluoromethyl)-1,4-phenylenediamine
009122-1G
009122-5G
FLUO009122-1GEA
0
GBP
FLUO009122-1G
FLUO009122-5G
α,α,α-Trifluoro-2,5-diaminotoluene
α,α,α-Trifluoro-2,5-diaminotoluene
Formula:
C₇H₇F₃N₂ MW: 176.14 Boiling Pt: 117…118 °C (7 mmHg) Melting Pt: 58 |
MDL Number:
MFCD00036101 CAS Number: 364-13-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...