1-Cycloheptylpiperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008914-5G
008914-10G
008914-1G
FLUO008914-5GEA
0
GBP
FLUO008914-5G
FLUO008914-10G
FLUO008914-1G
1-Cycloheptylpiperazine
1-Cycloheptylpiperazine
Formula:
C₁₁H₂₂N₂ |
MDL Number:
MFCD01076573 CAS Number: 21043-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...