2,4-Dichlorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,4-Dichlorophenylisothiocyanate, 1,3-Dichloro-4-isothiocyanatobenzene
008792-5G
008792-25G
FLUO008792-5GEA
0
GBP
FLUO008792-5G
FLUO008792-25G
2,4-Dichlorophenyl isothiocyanate
2,4-Dichlorophenyl isothiocyanate
Formula:
C₇H₃Cl₂NS Boiling Pt: 260 °C (1013 hPa) Melting Pt: 58…59 °C Density: 1.41 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037827 CAS Number: 6590-96-1 EINECS: 229-527-6 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...