3-Cyanophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Cyano-3-isothiocyanatobenzene, 3-Isothiocyanatobenzonitrile

008780-1G 008780-5G
FLUO008780-1GEA 0 GBP
FLUO008780-1G FLUO008780-5G
3-Cyanophenyl isothiocyanate
3-Cyanophenyl isothiocyanate
Formula: C₈H₄N₂S
MW: 160.2 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 63 °C
MDL Number: MFCD00041070
CAS Number: 3125-78-8
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR