5-Chloro-2-methoxyphenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008762-10G
008762-1G
008762-5G
FLUO008762-10GEA
0
GBP
FLUO008762-10G
FLUO008762-1G
FLUO008762-5G
5-Chloro-2-methoxyphenyl isothiocyanate
5-Chloro-2-methoxyphenyl isothiocyanate
Formula:
C₈H₆ClNOS Boiling Pt: 285 °C (1013 hPa) Melting Pt: 61 °C |
MDL Number:
MFCD00041073 CAS Number: 63429-99-2 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...