4-Isothiocyanatobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008758-5G
008758-1G
FLUO008758-5GEA
0
GBP
FLUO008758-5G
FLUO008758-1G
4-Isothiocyanatobenzoic acid
4-Isothiocyanatobenzoic acid
Formula:
C₈H₅NO₂S |
MDL Number:
MFCD00041369 CAS Number: 2131-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...